2-(Tributylstannyl)pyridine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Tri-n-butylstannyl)pyridine

OR3779-25G OR3779-5G OR3779-100G
77409-364EA 134.48 USD
77409-364 77409-366 77409-362
2-(Tributylstannyl)pyridine 95%
2-(Tributylstannyl)pyridine

Remainder tributyltin halide.

Formula: C₁₇H₃₁NSn
MW: 368.15
Boiling Pt: 132…134 °C/1 mmHg
Density: 1.137 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Freezer
MDL Number: MFCD00052052
CAS Number: 17997-47-6
UN: 2929
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR