5-Methylbenzo[b]thiophene

Supplier: APOLLO SCIENTIFIC
Warning

OR26010-1G OR26010-250MG
77407-728EA 29.2 USD
77407-728 77407-730
5-Methylbenzo[b]thiophene
5-Methylbenzo[b]thiophene
Formula: C₉H₈S
MW: 148.23
Boiling Pt: 71…73 °C/0.6 mmHg
Melting Pt: 32…37 °C
Density: 1.11 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052509
CAS Number: 14315-14-1
EINECS: 238-256-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR