5-Bromo-2-hydroxypyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR8599-1G OR8599-25G OR8599-5G
77424-554EA 21.26 USD
77424-554 77424-556 77424-558
5-Bromo-2-hydroxypyrimidine
5-Bromo-2-hydroxypyrimidine
Formula: C₄H₃BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD01318956
CAS Number: 38353-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR