2-Bromo-3-formylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde

OR6170-5G OR6170-100G OR6170-25G
77423-636EA 48.9 USD
77423-636 77423-632 77423-634
2-Bromo-3-formylpyridine 98%
2-Bromo-3-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Boiling Pt: 100 °C/3 mmHg
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD04966945
CAS Number: 128071-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR