Isonicotinaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

OR5048-500G OR5048-100G
77422-360EA 311.76 USD
77422-360 77422-354
Isonicotinaldehyde 98%
Isonicotinaldehyde
Formula: C₆H₅NO
Boiling Pt: 71…71 °C/10 mmHg
Melting Pt: –4 °C
Density: 1.137 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006425
CAS Number: 872-85-5
EINECS: 212-832-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR