3',5'-Bis(trifluoromethyl)propiophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)propan-1-one

PC3116-100G PC3116-25G PC3116-5G
77428-540EA 289.6 USD
77428-540 77428-544 77428-546
3',5'-Bis(trifluoromethyl)propiophenone 98%
3',5'-Bis(trifluoromethyl)propiophenone
Formula: C₁₁H₈F₆O
Boiling Pt: 72 °C/7 mmHg
Density: 1.379 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009911
CAS Number: 85068-34-4
EINECS: 285-296-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR