Coronene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Hexabenzobenzene

OR40595-1G OR40595-250MG
77421-182EA 207.84 USD
77421-182 77421-184
Coronene
Coronene
Formula: C₂₄H₁₂
MW: 300.36 g/mol
Boiling Pt: 525 °C
Melting Pt: 428 °C
Storage Temperature: Ambient
MDL Number: MFCD00004134
CAS Number: 191-07-1
EINECS: 205-881-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR