4,5-Difluoro-o-phenylenediamine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Difluoro-1,2-phenylenediamine, 4,5-Difluorobenzene-1,2-diamine, 1,2-Diamino-4,5-difluorobenzene

PC0342-100G PC0342-25G PC0342-5G
77426-220EA 330.1 USD
77426-220 77426-222 77426-224
4,5-Difluoro-o-phenylenediamine
4,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
Melting Pt: 131…132 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061131
CAS Number: 76179-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR