1-Bromo-3-fluoro-4-nitrobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-2-fluoronitrobenzene

PC0939-100G PC0939-25G PC0939-5G
77426-476EA 108 USD
77426-476 77426-478 77426-480
1-Bromo-3-fluoro-4-nitrobenzene 97%
1-Bromo-3-fluoro-4-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD01930221
CAS Number: 321-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR