5-Chloronicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR4770-1G OR4770-5G
77422-046EA 21.26 USD
77422-046 77422-048
5-Chloronicotinic acid
5-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00128901
CAS Number: 22620-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR