2'-Fluoro-4'-methoxyacetophenone 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC3817B-100G
PC3817B-25G
PC3817B-5G
77429-016EA
108
USD
77429-016
77429-018
77429-020
2'-Fluoro-4'-methoxyacetophenone 98%
2'-Fluoro-4'-methoxyacetophenone
Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).
Formula:
C₉H₉FO₂ Boiling Pt: 80…85 °C/0.1 mmHg Melting Pt: 51…53 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042290 CAS Number: 74457-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...