3-Chloro-4-methylphenyl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR23984-25G OR23984-5G OR23984-100G
77407-394EA 98.82 USD
77407-394 77407-396 77407-392
3-Chloro-4-methylphenyl isocyanate 97%
3-Chloro-4-methylphenyl isocyanate

  • May form small amount of sediment.

Formula: C₈H₆ClNO
Boiling Pt: 107 °C/3 mmHg
Melting Pt: 20…24 °C
Density: 1.224 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013858
CAS Number: 28479-22-3
EINECS: 249-050-7
UN: 2236
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR