2'-Bromo-4'-fluoroacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2-Bromo-4-fluorophenyl)ethanone

PC1396B-25G PC1396B-5G
77413-146EA 95.76 USD
77413-146 77413-148
2'-Bromo-4'-fluoroacetophenone 97%
2'-Bromo-4'-fluoroacetophenone
Formula: C₈H₆BrFO
Boiling Pt: 150 °C/12 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR