Isonicotinamide 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Pyridinecarboxylic acid amide

OR0914-100G OR0914-25G
77402-308EA 55.02 USD
77402-308 77402-310
Isonicotinamide 99%
Isonicotinamide
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR