2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

PC48678-25G PC48678-5G
77430-310EA 28.42 USD
77430-310 77430-312
2-Chloro-5-(trifluoromethyl)benzoic acid, Technical Grade
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR