2-(Trifluoromethoxy)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Trifluoromethoxy)benzene-1-sulphonyl chloride, 2-(Trifluoromethoxy)benzene-1-sulfonyl chloride

PC7436L-100G PC7436L-25G PC7436L-5G
77417-878EA 95.76 USD
77417-878 77417-880 77417-882
2-(Trifluoromethoxy)benzenesulfonyl chloride 97%
2-(Trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₃S
Boiling Pt: 75…77 °C/11 mmHg
Melting Pt: 27…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00052316
CAS Number: 103008-51-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR