5-Amino-2-fluorobenzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1071P-100G PC1071P-25G PC1071P-5G
77426-746EA 144.68 USD
77426-746 77426-748 77426-750
5-Amino-2-fluorobenzoic acid 97%
5-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00077449
CAS Number: 56741-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR