N-(1-Naphthyl)ethylenediamine dihydrochloride 92%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine
OR480629-1G
OR480629-25G
OR480629-5G
77422-148EA
21.26
USD
77422-148
77422-150
77422-152
N-(1-Naphthyl)ethylenediamine dihydrochloride 92%
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Formula:
C₁₂H₁₆Cl₂N₂ MW: 259.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012556 CAS Number: 1465-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...