p-Toluenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Toluenesulfonyl chloride, Toluene-p-sulfonyl chloride, Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-4-sulfonyl chloride, Toluene-p-sulphonyl chloride

OR9931-100G OR9931-1KG
77412-322EA 21.32 USD
77412-322 77412-324
p-Toluenesulfonyl chloride 97%
p-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 265
Melting Pt: 67
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8 II
Merck Index: 13,09612

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR