3-Bromo-2-hydroxypyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromopyridin-2(1H)-one, 3-Bromopyridin-2-ol, 3-Bromo-2-pyridinol

OR40206-1G OR40206-25G OR40206-5G
77421-048EA 47.88 USD
77421-048 77421-050 77421-052
3-Bromo-2-hydroxypyridine 98%
3-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 179…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411569
CAS Number: 13466-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR