3'-Fluoroacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3370-100G PC3370-25G PC3370-5G
77428-784EA 124.3 USD
77428-784 77428-786 77428-788
3'-Fluoroacetophenone 97%
3'-Fluoroacetophenone
Formula: C₈H₇FO
Boiling Pt: 81 °C/9 mmHg
Melting Pt: –3 °C
Density: 1.126 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000338
CAS Number: 455-36-7
EINECS: 207-245-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR