3,5-Difluorobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3,5-Difluorophenyl)methanol

PC2734-100G PC2734-10G PC2734-25G
77427-796EA 73.36 USD
77427-796 77427-798 77427-800
3,5-Difluorobenzyl alcohol 98%
3,5-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 98…99 °C/9 mmHg
Density: 1.276 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00010313
CAS Number: 79538-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR