Resveratrol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

OR46018-100G OR46018-1KG OR46018-500G
77421-746EA 93.74 USD
77421-746 77421-748 77421-750
Resveratrol 99%
Resveratrol
Formula: C₁₄H₁₂O₃
Melting Pt: 261…267 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00133799
CAS Number: 501-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR