5,6-Dimethoxyindole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5372-1G OR5372-250MG
77422-840EA 96.78 USD
77422-840 77422-842
5,6-Dimethoxyindole 98%
5,6-Dimethoxyindole
Formula: C₁₀H₁₁NO₂
Boiling Pt: 198 °C/8 mmHg
Melting Pt: 154…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005675
CAS Number: 14430-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR