1,8-Dihydroxy-9,10-anthraquinone 98+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Dantron, Danthron, 1,8-Dihydroxyanthraquinone

OR61434-100MG OR61434-10G OR61434-25G
77423-588EA 21.26 USD
77423-588 77423-590 77423-592
1,8-Dihydroxy-9,10-anthraquinone 98+%
1,8-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Melting Pt: 190…195 °C
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00001211
CAS Number: 117-10-2
EINECS: 204-173-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR