1,4-Dibromonaphthalene 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR30594-25G OR30594-5G
77408-902EA 39 USD
77408-902 77408-904
1,4-Dibromonaphthalene 98%
1,4-Dibromonaphthalene
Formula: C₁₀H₆Br₂
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00041823
CAS Number: 83-53-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR