2-Nitrophenol
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR01570-25G
OR01570-5G
77401-768EA
44.82
USD
77401-768
77401-770
2-Nitrophenol
2-Nitrophenol
Formula:
C₆H₅NO₃ Boiling Pt: 214…216 °C Melting Pt: 44…48 °C Density: 1.26 g/cm³ (25 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011688 CAS Number: 88-75-5 UN: 1663 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...