Aniline 99+% ACS
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Aminobenzene, Phenylamine
OR43506-100ML
OR43506-1L
OR43506-500ML
77409-692EA
89.66
USD
77409-692
77409-694
77409-696
Aniline 99+% ACS
Aniline
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 184 Melting Pt: -6.2 Density: 1.022 g/cm³ (25 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1 II Merck Index: 13,00661 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...