3,4,5-Trifluoroanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,2,3-Trifluoro-5-methoxybenzene, Trifluoro-3,4,5-anisole, 1-Methoxy-3,4,5-trifluorobenzene

PC0513-10G PC0513-25G PC0513-5G
77412-602EA 118.36 USD
77412-602 77412-604 77412-606
3,4,5-Trifluoroanisole
3,4,5-Trifluoroanisole
Formula: C₇H₅F₃O
MW: 162.11
Boiling Pt: 39 °C/1 mmHg
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00042397
CAS Number: 203245-17-4
UN: 3271
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR