1,2-Diiodotetrafluorobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2969-1G PC2969-25G PC2969-5G
77428-052EA 34.64 USD
77428-052 77428-054 77428-056
1,2-Diiodotetrafluorobenzene 97%
1,2-Diiodotetrafluorobenzene
Formula: C₆F₄I₂
MW: 401.87 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019011
CAS Number: 2708-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR