2-Chloro-5-(trifluoromethyl)benzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5613-100G PC5613-25G PC5613-5G
77431-774EA 151.8 USD
77431-774 77431-776 77431-778
2-Chloro-5-(trifluoromethyl)benzaldehyde 97%
2-Chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Boiling Pt: 42…44 °C/1.5 mmHg
Density: 1.435 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00134430
CAS Number: 82386-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR