4-(Trifluoromethoxy)-1,2-phenylenediamine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethoxy)benzene-1,2-diamine

PC5195-1G PC5195-25G PC5195-5G
77431-176EA 24.46 USD
77431-176 77431-178 77431-180
4-(Trifluoromethoxy)-1,2-phenylenediamine 97%
4-(Trifluoromethoxy)-1,2-phenylenediamine
Formula: C₇H₇F₃N₂O
MW: 192.14 g/mol
Boiling Pt: 106…108 °C/5 mmHg
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094278
CAS Number: 658-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR