3-(3-Trifluoromethylphenyl)propionic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid

PC5441-100G PC5441-25G PC5441-5G
77431-648EA 219.04 USD
77431-648 77431-650 77431-652
3-(3-Trifluoromethylphenyl)propionic acid 97%
3-(3-Trifluoromethylphenyl)propionic acid
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Boiling Pt: 149 °C/11 mmHg
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00090487
CAS Number: 585-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR