Palladium(II) trifluoroacetate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

PC5367-5G PC5367-1G
77431-616EA 360.66 USD
77431-616 77431-614
Palladium(II) trifluoroacetate 98%
Palladium (II) trifluoroacetate
Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR