5-Chloro-2-(trifluoromethyl)aniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-4-chlorobenzotrifluoride

PC49064-10G PC49064-1G PC49064-5G
77430-360EA 146.7 USD
77430-360 77430-362 77430-364
5-Chloro-2-(trifluoromethyl)aniline 95%
5-Chloro-2-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MW: 195.5700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08282771
CAS Number: 445-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR