Methyl-2-fluorobenzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-fluorobenzoate, 2-Fluorobenzoic acid emthyl ester

PC5088-100G PC5088-250G PC5088-25G
77431-026EA 41.78 USD
77431-026 77431-028 77431-030
Methyl-2-fluorobenzoate 98%
Methyl 2-fluorobenzoate
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 109…110 °C/35 mmHg
Density: 1.21 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00017913
CAS Number: 394-35-4
EINECS: 206-894-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR