2-Cyclopropyl-5-fluoroaniline
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC49423-1G
PC49423-250MG
PC49423-5G
77430-560EA
73.36
USD
77430-560
77430-562
77430-564
2-Cyclopropyl-5-fluoroaniline
2-Cyclopropyl-5-fluoroaniline
Formula:
C₉H₁₀FN MW: 151.181 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD30322597 CAS Number: 1538364-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...