2-Hydroxy-3-(trifluoromethyl)pyridine 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3-(Trifluoromethyl)pyridin-2-ol
PC4868W-100G
PC4868W-10G
PC4868W-25G
77430-314EA
73.36
USD
77430-314
77430-316
77430-318
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-(trifluoromethyl)pyridine
Formula:
C₆H₄F₃NO MW: 163.1 Melting Pt: 150…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153194 CAS Number: 22245-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...