1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea

Supplier: APOLLO SCIENTIFIC
Warning

PC48673-1G PC48673-250MG
77430-298EA 196.64 USD
77430-298 77430-300
1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea
1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea
Formula: C₉H₉BrClFN₂O
MW: 295.536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31372515
CAS Number: 2089312-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR