(Nonafluorobutyl)undecafluorocyclohexane

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluoro(butylcyclohexane)

PC408080-1G PC408080-25G PC408080-5G
77429-372EA 21.26 USD
77429-372 77429-374 77429-376
(Nonafluorobutyl)undecafluorocyclohexane
(Nonafluorobutyl)undecafluorocyclohexane
Formula: C₁₀F₂₀
MW: 500.08
Boiling Pt: 145…147 °C
Density: 1.899 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 374-60-7
EINECS: 206-777-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR