1,4-Bis(trimethylsilyl)tetrafluorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

PC450555-100G PC450555-25G
77429-956EA 687.7 USD
77429-956 77429-958
1,4-Bis(trimethylsilyl)tetrafluorobenzene
1,4-Bis(trimethylsilyl)tetrafluorobenzene
Formula: C₁₂H₁₈F₄Si₂
MW: 294.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00093109
CAS Number: 16956-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR