2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC450217-250MG PC450217-100MG PC450217-1G
77429-896EA 205.6 USD
77429-896 77429-892 77429-894
2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether 97%
2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether
Formula: C₆H₃F₁₁O
MW: 300.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27918475
CAS Number: 1661005-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR