5-Fluoro-3-methylindole

Supplier: APOLLO SCIENTIFIC
Warning

PC3027-1G PC3027-250MG
77428-318EA 206.82 USD
77428-318 77428-320
5-Fluoro-3-methylindole
5-Fluoro-3-methylindole
Formula: C₉H₈FN
Melting Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD03095377
CAS Number: 392-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR