1,2-Difluoro-4-iodobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4-Difluoroiodobenzene

PC2836-100G PC2836-25G
77427-910EA 79.46 USD
77427-910 77427-912
1,2-Difluoro-4-iodobenzene 99%
1,2-Difluoro-4-iodobenzene
Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 177
Density: 1.99 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044630
CAS Number: 64248-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR