1-Bromo-3-chloro-5-fluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Chloro-5-fluorobromobenzene

PC1367M-100G PC1367M-25G PC1367M-5G
77426-976EA 54 USD
77426-976 77426-978 77426-980
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene
Formula: C₆H₃BrClF
Density: 1.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070747
CAS Number: 33863-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR