7-Amino-4-(trifluoromethyl)coumarin 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC1100CF-10G PC1100CF-1G PC1100CF-5G
77426-800EA 134.48 USD
77426-800 77426-802 77426-804
7-Amino-4-(trifluoromethyl)coumarin 99%
7-Amino-4-(trifluoromethyl)coumarin
Formula: C₁₀H₆F₃NO₂
Melting Pt: 221…223 °C
Density: 1.59 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006858
CAS Number: 53518-15-3
EINECS: 258-599-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR