4-Fluoro-3-methylbenzamide

Supplier: APOLLO SCIENTIFIC

PC0344-10G PC0344-1G PC0344-5G
77426-226EA 267.94 USD
77426-226 77426-228 77426-230
4-Fluoro-3-methylbenzamide
4-Fluoro-3-methylbenzamide
Formula: C₈H₈FNO
MW: 153.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631489
CAS Number: 261945-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR