(Bis(trifluoroacetoxy)iodo)benzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [Bis(trifluoroacetoxy)](phenyl)iodane

PC0403-100G PC0403-25G
77426-238EA 83.54 USD
77426-238 77426-240
(Bis(trifluoroacetoxy)iodo)benzene 97%
(Bis(trifluoroacetoxy)iodo)benzene

Used to estimate asparagine and glutamine in peptides and proteins.

Formula: C₁₀H₅F₆IO₄
Melting Pt: 122…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009672
CAS Number: 2712-78-9
EINECS: 220-308-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR