1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4'-Amino-2,2,2-trifluoroacetophenone, 1-(4-Aminophenyl)-2,2,2-trifluoroethanone

PC10234-1G PC10234-5G
77426-546EA 123.28 USD
77426-546 77426-548
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone
Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663790
CAS Number: 23516-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR