2-Amino-5-fluoropyridine 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC0886-10G PC0886-1G PC0886-5G
77426-448EA 44.82 USD
77426-448 77426-450 77426-452
2-Amino-5-fluoropyridine 99%
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11
Melting Pt: 93…99 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861120
CAS Number: 21717-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR